Stampa…

3-Benzyloxyaniline ≥98%

Fornitore: Thermo Scientific
Attenzione

H55708.06 H55708.14
H55708.06EA 72.5 EUR
H55708.06 H55708.14
3-Benzyloxyaniline ≥98%
3-Benzyloxyaniline

Purity: 98% CAS No.: 1484-26-0 Molecular Formula: C13H13NO Formula Weight: 199.25 Physical Form: Solid Appearance: Pale brown Melting Point: 63-67° MDL No.: MFCD00007784

Formula: C₁₃H₁₃NO
Peso molecolare: 199,25 g/mol
Punto di fusione: 63…67 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007784
CAS Number: 1484-26-0
EINECS: 216-056-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR