Stampa…
Washers, BOLA
Fornitore: Bohlender
H1126-26 H1126-22 H1126-18 H1126-14
BOHLH1126-26EA 3 EUR
BOHLH1126-26 BOHLH1126-22 BOHLH1126-18 BOHLH1126-14
Washers, BOLA
Guarnizioni Anelli di tenuta
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 125-1