Stampa…
Safety helmets, uvex airwing B
Fornitore: UVEX
9762.020 9762.120 9762.220 9762.320 9762.420 9762.520
111-8232EA 16.7 EUR
111-8232 111-8235 111-8216 111-8217 111-8218 111-8219
Safety helmets, uvex airwing B
Elmetti Elmetti di sicurezza
Helmet shell made from polyethylene (PE).
  • Three variable ventilation openings
  • Conventional interior design ensuring comfortable fit
Size: 510 to 610 mm.
Peso: 360 g.
Certificazioni: EN 397