Stampa…

2-Amino-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A0874-25G A0874-5G
TCIAA0874-25GEA 122 EUR
TCIAA0874-25G TCIAA0874-5G
2-Amino-5-methyl-1,3,4-thiadiazole ≥98.0% (by titrimetric analysis)
2-Amino-5-methyl-1,3,4-thiadiazole
Formula: C₃H₅N₃S
Peso molecolare: 115,16 g/mol
MDL Number: MFCD00003110
CAS Number: 108-33-8
EINECS: 203-573-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ffba6e62ed8aa30f52ba47ab0c8f2dcec092fb7&vid=6ffba6e62ed8aa30f52ba47ab0c8f2dcec092fb7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR