Stampa…

Bismut(III) gallate basic hydrate

Fornitore: Thermo Scientific

Synonyms: Bismuth subgallate hydrate, 2,7-Dihydroxy-1,3,2-benzodioxabismole-5-carboxylic acid hydrate, Dermatol hydrate

B24187.22 B24187.36
B24187.22EA 35.3 EUR
B24187.22 B24187.36
Bismut(III) gallate basic hydrate
Bismut(III) gallate basic hydrate

CAS No.: 99-26-3 Molecular Formula: C7H5BiO6.xH2O Formula Weight: 394.10(anhy) MDL No.: MFCD01632692

Formula: C₇H₅BiO₆·xH₂O
Peso molecolare: 394,09 g/mol (anhy)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01632692
CAS Number: 342406-26-2
EINECS: 202-742-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR