Stampa…

Sulphathiazole ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: 4-Amino-N-(thiazol-2-yl)benzenesulphonamide, N1-2-Thiazolylsulphanilamid

A10727.22 A10727.36
A10727.22EA 73 EUR
A10727.22 A10727.36
Sulphathiazole ≥99%
Sulfathiazole

Purity: 99% CAS No.: 72-14-0 Molecular Formula: C9H9N3O2S2 Formula Weight: 255.32 Melting Point: 200-203° Merck Reference: 14,8943 MDL No.: MFCD00005319


Short-acting antimicrobial topical sulfa drug

Formula: C₉H₉N₃O₂S₂
Peso molecolare: 255.32 g/mol
Punto di fusione: 200…203 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005319
CAS Number: 72-14-0
EINECS: 200-771-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR