DL-Panthenol ≥99%
Fornitore: Thermo Fisher Scientific
Purity: 99%
CAS No.: 16485-10-2
Molecular Formula: C9H19NO4
Formula Weight: 205.26
Storage Temperature: 2°C to 8°C
Melting Point: 66-69°
MDL No.: MFCD00002944
Formula:
C₉H₁₉NO₄ Punto di fusione: 66…69 °C |
MDL Number:
MFCD00002944 CAS Number: 16485-10-2 EINECS: 240-540-6 Merck Index: 14,2947 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...