5-Decyne ≥98%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 98%
CAS No.: 1942-46-7
Molecular Formula: CH3(CH2)3C≡C(CH2)3CH3
Formula Weight: 138.26
Flash Point: 54°(129°F)
Refractive Index: 1.4330
Odour: Petrol-like
Boiling Point: 177-178°
Beilstein Reference: 1697840
UN No.: 3295
MDL No.: MFCD00027277
Formula:
C₁₀H₁₈ Punto di ebollizione: 177…178 °C Densità: 0,770 Flash Pt: 54 °C (129 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00027277 CAS Number: 1942-46-7 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...