Methyl isobutyrylacetate per sintesi, Sigma-Aldrich®
Fornitore: Merck
Synonyms:
Methyl 4-methyl-3-oxopentanoate, Methyl 4-methyl-3-oxovalerate
M-Clarity™ quality level = MQ200
Formula:
C₇H₁₂O₃ Peso molecolare: 144.17 g/mol |
MDL Number:
MFCD00040499 CAS Number: 42558-54-3 EINECS: 418-900-0 |
Risultati del test di specificazione
Solubility | 43.6 g/l (20 °C) |
Melting Point | -75 °C |
Molar Mass | 144.17 g/mol |
Boiling Point | 192 °C (1013 hPa) |
Vapor Pressure | 1.3 hPa (20 °C) |
Flash Point | 79 °C |
Explosion Limit | 1.29 %(V) |
Density | 1.01 g/cm3 (20 °C) |
pH | 4.8 (10 g/l, H2O, 20 °C) |
Ignition Point | 280 °C |
Synonyms | Isobutyrylacetic acid methyl ester, Methyl 4-methyl-3-oxopentanoate |
Hill Formula | C7H12O3 |
Chemical Formula | CH3CH(CH3)COCH2COOCH3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...