Stampa…

Benzo[b]thiophene ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Thianaphthene, 1-Benzothiophene

B0093-25G B0093-5G
TCIAB0093-25GEA 54 EUR
TCIAB0093-25G TCIAB0093-5G
Benzo[b]thiophene ≥97.0% (by GC)
Benzo[b]thiophene
Formula: C₈H₆S
Peso molecolare: 134,2 g/mol
Punto di ebollizione: 221°C
MDL Number: MFCD00005864
CAS Number: 95-15-8
EINECS: 202-395-7
UN: 3077
ADR: 9,III
Merck Index: 14,09303
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f804e07394d69dbbc14ed2a6d07b62572997f60&vid=2f804e07394d69dbbc14ed2a6d07b62572997f60&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow to Light orange powder to lump to clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR