Stampa…

Bis(aminoguanidinium)sulphate per sintesi, Sigma-Aldrich®

Fornitore: Merck


M-Clarity™ quality level = MQ200

8.20073.9015 8.20073.9050
8.20073.9015EA 0 EUR
8.20073.9015 8.20073.9050
Bis(aminoguanidinium)sulphate per sintesi, Sigma-Aldrich®
Bis(aminoguanidinium)sulphate
Formula: C₂H₁₄N₈O₄S
MDL Number: MFCD00035665
CAS Number: 996-19-0
EINECS: 213-628-7

Visualizza i codici

Risultati del test di specificazione

Solubility 170 g/l (20 °C)
Melting Point 208 - 210 °C
Molar Mass 246.24 g/mol
Bulk Density 750 kg/m3
Density 1.36 g/cm3 (20 °C)
pH 4 - 6 (25 g/l, H2O, 20 °C)

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR