4-Guanidinobenzoic acid hydrochloride ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
N-(4-Carboxyphenyl)guanidine hydrochloride
G0238-25G
G0238-5G
TCIAG0238-25GEA
87
EUR
TCIAG0238-25G
TCIAG0238-5G
4-Guanidinobenzoic acid hydrochloride ≥97.0% (by titrimetric analysis)
4-Guanidinobenzoic acid hydrochloride
Formula:
C₈H₁₀ClN₃O₂ Peso molecolare: 215,64 g/mol |
MDL Number:
MFCD00040587 CAS Number: 42823-46-1 EINECS: 255-956-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...