Stampa…

(±)-Ethyl-2-bromopropionate ≥98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate

A10820.18 A10820.30 A10820.36
A10820.18EA 23.4 EUR
A10820.18 A10820.30 A10820.36
(±)-Ethyl-2-bromopropionate ≥98%
(±)-Ethyl-2-bromopropionate

Purity: 98+% CAS No.: 535-11-5 Molecular Formula: C5H9BrO2 Formula Weight: 181.03 Flash Point: 51°(123°F) Refractive Index: 1.4460 Boiling Point: 158-160° Beilstein Reference: 773920 Merck Reference: 14,3772 UN No.: 1992 MDL No.: MFCD00000144

Formula: C₅H₉BrO₂
Peso molecolare: 181,03 g/mol
Punto di ebollizione: 158…160 °C
Densità: 1,391
Flash Pt: 51 °C (123 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-5
UN: 1992
ADR: 3,III
Merck Index: 14,3772

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR