2,3,5-Trimethylthiophene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
T1327-1G
T1327-5G
TCIAT1327-1GEA
84
EUR
TCIAT1327-1G
TCIAT1327-5G
2,3,5-Trimethylthiophene ≥98.0% (by GC)
2,3,5-Trimethylthiophene
Formula:
C₇H₁₀S Punto di ebollizione: 82°C |
MDL Number:
MFCD00142784 CAS Number: 1795-05-7 EINECS: 217-269-7 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...