Stampa…
Piston rods for Transferpettor
Fornitore: Brand
701928 701932 701934 701938 701936 701930
612-5330EA 39.5 EUR
612-5330 612-3349 BRND701934 BRND701938 BRND701936 BRND701930
Piston rods for Transferpettor
Pipette Pipette automatiche
For capacities ranged from 1 to 200 µl, provided with seal.
Informazioni per l'ordine: Pack of 3.