Zinco ossido, RonaCare®
Fornitore: Merck
|
Attenzione
|
Synonyms:
Zinci oxidum, Zinc (II) oxide
Formula:
ZnO Peso molecolare: 81.39 g/mol Densità: 5.61 g/cm³ (25 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9 III REACH: 01-2119463881-32 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...