Stampa…

Ammonium perrhenate ≥99.999% (metals basis), Puratronic®

Fornitore: Thermo Scientific
Pericolo

10846.04 10846.09
10846.04EA 132 EUR
10846.04 10846.09
Ammonium perrhenate ≥99.999% (metals basis), Puratronic®
Ammonium perrhenate

Purity: 99.999% CAS No.: 13598-65-7 Molecular Formula: NH4ReO4 Formula Weight: 268.24 Physical Form: Crystalline Density: 3.97 Appearance: White Melting Point: dec. UN No.: 1479 MDL No.: MFCD00012377

Formula: H₄NO₄Re
Peso molecolare: 268,24 g/mol
Punto di fusione: Dec.
Densità: 3,97
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012377
CAS Number: 13598-65-7
EINECS: 237-075-6
UN: 1479
ADR: 5.1,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR