Stampa…

Phenylacetamide ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: 2-Phenylacetamide

P0120-25G P0120-500G
TCIAP0120-25GEA 29 EUR
TCIAP0120-25G TCIAP0120-500G
Phenylacetamide ≥98.0% (by total nitrogen basis)
Phenylacetamide
Formula: C₈H₉NO
Peso molecolare: 135,17 g/mol
MDL Number: MFCD00059193
CAS Number: 103-81-1
EINECS: 203-147-0
Merck Index: 14,07266
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=164040ad4351aae3c277498adbc5e90eb0561a72&vid=164040ad4351aae3c277498adbc5e90eb0561a72&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 156.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR