Stampa…

Sodium methanesulphinate ≥90.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

M1255-25G M1255-5G
TCIAM1255-25GEA 144 EUR
TCIAM1255-25G TCIAM1255-5G
Sodium methanesulphinate ≥90.0% (by titrimetric analysis)
Sodium methanesulphinate
Formula: CH₃NaO₂S
MDL Number: MFCD00040392
CAS Number: 20277-69-4
EINECS: 243-669-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd8ea49391faa5e53dc3c0402746e8c304a5d428&vid=cd8ea49391faa5e53dc3c0402746e8c304a5d428&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR