4-Chloroisatin ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
4-Chloroindoline-2,3-dione
C1870-25G
C1870-5G
TCIAC1870-25GEA
113
EUR
TCIAC1870-25G
TCIAC1870-5G
4-Chloroisatin ≥97.0% (by GC, titration analysis)
4-Chloroisatin
Formula:
C₈H₄ClNO₂ Peso molecolare: 181,58 g/mol Punto di fusione: 258°C |
MDL Number:
MFCD00129178 CAS Number: 6344-05-4 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...