Stampa…

4-Chloropicolinic acid ≥96.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

C1360-1G C1360 C1360-5G
TCIAC1360-1GEA 65 EUR
TCIAC1360-1G FLUOC1360 TCIAC1360-5G
4-Chloropicolinic acid ≥96.0% (by GC, titration analysis)
4-Chloropicolinic acid
Formula: C₆H₄ClNO₂
MDL Number: MFCD00191400
CAS Number: 5470-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6676fb5bf3d83cc7c44c6632faf7b7a26d0c037b&vid=6676fb5bf3d83cc7c44c6632faf7b7a26d0c037b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR