Phenylphosphonic dichloride, Sigma-Aldrich®
Fornitore: Merck
|
Pericolo
|
Synonyms:
Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide
M-Clarity™ quality level = MQ200
Formula:
C₆H₅Cl₂OP Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002070 CAS Number: 824-72-6 EINECS: 212-534-3 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Solubility | (20 °C) (decomposition) |
Melting Point | 1 °C |
Molar Mass | 194.98 g/mol |
Boiling Point | 258 °C |
Flash Point | 204 °C |
Refractive Index | 1.5581 (25 °C, 589 nm) |
Density | 1.40 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...