Stampa…

Tetraethylene glycol, Sigma-Aldrich®

Fornitore: Merck

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol

M-Clarity™ quality level = MQ200

8.08619.1000 8.08619.9050
8.08619.1000EA 63.2 EUR
8.08619.1000 8.08619.9050
Tetraethylene glycol, Sigma-Aldrich®
Tetraethylene glycol
Formula: C₈H₁₈O₅
MDL Number: MFCD00002879
CAS Number: 112-60-7
EINECS: 203-989-9

Visualizza i codici

Risultati del test di specificazione

Solubility 1000 g/l (20 °C)
Melting Point -5 °C
Molar Mass 194.22 g/mol
Boiling Point 327 - 328 °C (1013 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 174 °C
Refractive Index 1.4609 (20 °C, 589 nm)
Explosion Limit 0.5 - 3.4 %(V)
Density 1.12 g/cm3 (20 °C)
pH 8.5 - 9.0 (500 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR