Stampa…

Anthrone, dark yellow powder

Fornitore: MP Biomedicals
Attenzione

Synonyms: Anthracen-9(10H)-one

0210069180 0210069110 0210069125
ICNA0210069180EA 358 EUR
ICNA0210069180 ICNA0210069110 ICNA0210069125
Anthrone, dark yellow powder
Anthrone

Anthrone is a tricyclic aromatic hydrocarbon.


It is used to determine hydrocarbons using a colorimetric method. It helps to estimate carbohydrates in yeast and plant extracts.


Anthrone is a tricyclic aromatic hydrocarbon.


It is used to determine hydrocarbons using a colorimetric method. It helps to estimate carbohydrates in yeast and plant extracts.


Soluble in ethyl acetate.


Attenzione: Protect from air oxidation

Formula: C₁₄H₁₀O
Peso molecolare: 194,23 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR