Stampa…

Methyl-5-hydroxynicotinate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

H0841-1G H0841-25G H0841-5G
TCIAH0841-1GEA 41 EUR
TCIAH0841-1G TCIAH0841-25G TCIAH0841-5G
Methyl-5-hydroxynicotinate ≥98.0% (by GC, titration analysis)
Methyl-5-hydroxynicotinate
Formula: C₇H₇NO₃
Peso molecolare: 153,14 g/mol
MDL Number: MFCD00191521
CAS Number: 30766-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1036875bbc12d524975b5f0972c328ffb2da7845&vid=1036875bbc12d524975b5f0972c328ffb2da7845&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 185.0 to 195.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR