Stampa…

Phthalamic acid ≥99%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 2-Carbamoylbenzoic acid

L12432.06 L12432.14
L12432.06EA 52.5 EUR
L12432.06 L12432.14
Phthalamic acid ≥99%
Phthalamic acid

Purity: 99% CAS No.: 88-97-1 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Melting Point: 140-143° Beilstein Reference: 1868205

Formula: C₈H₇NO₃
Peso molecolare: 165,15 g/mol
Punto di fusione: 140…143 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00025476
CAS Number: 88-97-1
EINECS: 201-871-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR