Benzylthiourea ≥98%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
1-Benzyl-2-thiourea, N-Benzylthiourea
Purity: 98%
CAS No.: 621-83-0
Molecular Formula: C8H10N2S
Formula Weight: 166.25
Odour: Odourless
Melting Point: 162-166°
Beilstein Reference: 972372
UN No.: 2811
MDL No.: MFCD00041370
Formula:
C₈H₁₀N₂S Punto di fusione: 162…166 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00041370 CAS Number: 621-83-0 EINECS: 210-709-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...