Stampa…

Tetraethylene glycol monomethyl ether ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: mPEG4-OH

A17021.14 A17021.06 A17021.22
A17021.14EA 226 EUR
A17021.14 A17021.06 A17021.22
Tetraethylene glycol monomethyl ether ≥98%
Tetraethylene glycol monomethyl ether

Purity: 98% CAS No.: 23783-42-8 Molecular Formula: H(OCH2CH2)4OCH3 Formula Weight: 208.25 Refractive Index: 1.4450 Boiling Point: 158-160°/5mm MDL No.: MFCD00041756

Formula: C₉H₂₀O₅
Peso molecolare: 208,25 g/mol
Punto di ebollizione: 158…160 °C (5 mmHg)
Densità: 1,07
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00041756
CAS Number: 23783-42-8
EINECS: 245-883-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR