Stampa…

Ethylenediammonium dichloride 98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Ethylenediamine dihydrochloride

A13563.0E A13563.22 A13563.36
A13563.0EEA 412 EUR
A13563.0E A13563.22 A13563.36
Ethylenediammonium dichloride 98%
Ethylenediammonium dichloride

Purity: 98% CAS No.: 333-18-6 Molecular Formula: HCl.H2NCH2CH2NH2.HCl Formula Weight: 133.02 Melting Point: 300° MDL No.: MFCD00012524

Formula: C₂H₁₀Cl₂N₂
Peso molecolare: 133,02 g/mol
Punto di fusione: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012524
CAS Number: 333-18-6
EINECS: 206-369-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR