Triethanolamine hydrochloride 99+%
Fornitore: Thermo Scientific
170051000
170055000
170050025
ACRO170051000EA
40.4
EUR
ACRO170051000
ACRO170055000
ACRO170050025
Triethanolamine hydrochloride 99+%
Triethanolamine hydrochloride
Formula:
C₆H₁₆ClNO₃ Peso molecolare: 185,65 g/mol Punto di fusione: 177…179 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00012596 CAS Number: 637-39-8 EINECS: 211-284-2 |
Risultati del test di specificazione
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥99.0 % |
Loss on drying | ≤1.0 % (105°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...