Aminoacetaldehyde dimethyl acetal 99%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
153340250
153341000
153345000
153340025
ACRO153340250EA
43.4
EUR
ACRO153340250
ACRO153341000
ACRO153345000
ACRO153340025
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal
Formula:
C₄H₁₁NO₂ Punto di ebollizione: 135…139 °C (95 mmHg) Densità: 0,97 g/cm³ Flash Pt: 41 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008135 CAS Number: 22483-09-6 EINECS: 245-026-5 UN: 2733 ADR: 3,III |
Risultati del test di specificazione
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4160 to 1.4180 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...