Stampa…

Diphenyliodonium chloride 97%

Fornitore: Thermo Scientific
Pericolo

117330100 117330500
ACRO117330100EA 188 EUR
ACRO117330100 ACRO117330500
Diphenyliodonium chloride 97%
Diphenyliodonium chloride
Formula: C₁₂H₁₀ClI
Peso molecolare: 316,57 g/mol
Punto di fusione: 227…228 °C (subl.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00011909
CAS Number: 1483-72-3
EINECS: 216-049-8
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
Chloride and Bromide ≤0.02 % (as Cl)
Iodide (I) passes test
Sulfate (SO4) ≤150 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %
Nitrogen compounds (as N) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR