Stampa…

Sodium perrhenate ≥99.95% (metals basis)

Fornitore: Thermo Scientific
Pericolo

62107.01 62107.03 62107.06
62107.01EA 40.4 EUR
62107.01 62107.03 62107.06
Sodium perrhenate ≥99.95% (metals basis)
Sodium perrhenate

Purity: 99.95% CAS No.: 13472-33-8 Molecular Formula: NaO4Re Formula Weight: 273.19 Physical Form: Crystalline Density: 5.390 Appearance: White Odour: Odourless Melting Point: 300° UN No.: 1479 MDL No.: MFCD00014231

Formula: NaO₄Re
Peso molecolare: 273,19 g/mol
Punto di fusione: 300 °C
Densità: 5,390
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014231
CAS Number: 13472-33-8
EINECS: 236-742-9
UN: 1479
ADR: 5.1,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR