Sodium perrhenate (68% Re) ≥99% (metals basis)
Fornitore: Thermo Scientific
|
Pericolo
|
11412.04
11412.09
11412.18
11412.04EA
62.6
EUR
11412.04
11412.09
11412.18
Sodium perrhenate (68% Re) ≥99% (metals basis)
Sodium perrhenate
Purity: 99+%
CAS No.: 13472-33-8
Molecular Formula: NaO4Re
Formula Weight: 273.19
Physical Form: Crystalline
Density: 5.390
Appearance: White
Odour: Odourless
Melting Point: 300°
UN No.: 1479
MDL No.: MFCD00014231
Formula:
NaO₄Re Peso molecolare: 273,19 g/mol Punto di fusione: 300 °C Densità: 5,390 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00014231 CAS Number: 13472-33-8 EINECS: 236-742-9 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...