Stampa…

Trilithium orthophosphate ≥99.99% (metals basis), Puratronic®

Fornitore: Thermo Scientific
Attenzione

Synonyms: Lithium phosphate tribasic, Lithium orthophosphate, Lithium phosphate

10743.14 10743.22
10743.14EA 179 EUR
10743.14 10743.22
Trilithium orthophosphate ≥99.99% (metals basis), Puratronic®
Trilithium orthophosphate

Purity: 99.99% CAS No.: 10377-52-3 Molecular Formula: Li3PO4 Formula Weight: 115.79 Physical Form: Powder Density: 2.54 Appearance: White Odour: Odourless Melting Point: 837° MDL No.: MFCD00016187

Formula: Li₃O₄P
Peso molecolare: 115,79 g/mol
Punto di fusione: 837 °C
Densità: 2,54
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016187
CAS Number: 10377-52-3
EINECS: 233-823-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR