4-Bromophthalic anhydride ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
5-Bromoisobenzofuran-1,3-dione
Formula:
C₈H₃BrO₃ Punto di ebollizione: 301°C |
MDL Number:
MFCD00191323 CAS Number: 86-90-8 EINECS: 201-707-9 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Morpholine Method) | min. 95.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...