3-Methylcrotonic acid
Fornitore: Merck
Formula:
C₅H₈O₂ |
MDL Number:
MFCD00004366 CAS Number: 541-47-9 EINECS: 208-782-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...