Stampa…
Methyl-2,6-dichlorobenzoate 97%
Fornitore: Thermo Scientific
Synonyms: Methyl 2,6-dichlorobenzoate
B24912.03 B24912.06 B24912.14
B24912.03EA 24.7 EUR
B24912.03 B24912.06 B24912.14
Methyl-2,6-dichlorobenzoate 97%
Methyl-2,6-dichlorobenzoate
Formula: C₈H₆Cl₂O₂
Peso molecolare: 205,04 g/mol
MDL Number: MFCD01313701
CAS Number: 14920-87-7