Stampa…

p-Toluic hydrazide ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: p-Toluoylhydrazine, 4-Methylbenzohydrazide, p-Toluic acid hydrazide, p-Toluic Hydrazide

A12604.06 A12604.14
A12604.06EA 23.9 EUR
A12604.06 A12604.14
p-Toluic hydrazide ≥98%
p-Toluic hydrazide

Purity: 98% CAS No.: 3619-22-5 Molecular Formula: C8H10N2O Formula Weight: 150.18 Melting Point: 115-118° Beilstein Reference: 607602

Formula: C₈H₁₀N₂O
Peso molecolare: 150,18 g/mol
Punto di fusione: 115…118 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007607
CAS Number: 3619-22-5
EINECS: 222-815-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR