Iodocyclohexane ≥98% stabilizzato
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Stabilization:
Stabilised with copper
Stabilized with copper
Purity: 98%
CAS No.: 626-62-0
Molecular Formula: C6H11I
Formula Weight: 210.06
Flash Point: 71°(159°F)
Refractive Index: 1.5475
Boiling Point: 180°
Beilstein Reference: 1900797
MDL No.: MFCD00003826
Formula:
C₆H₁₁I Punto di ebollizione: 180 °C Densità: 1,625 Flash Pt: 71 °C (159 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003826 CAS Number: 626-62-0 EINECS: 210-957-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...