7-Methyl-8-nitroquinoline ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
L03882.06
L03882.14
L03882.06EA
109
EUR
L03882.06
L03882.14
7-Methyl-8-nitroquinoline ≥98%
7-Methyl-8-nitroquinoline
Purity: 98+%
CAS No.: 7471-63-8
Molecular Formula: C10H8N2O2
Formula Weight: 188.19
Melting Point: 185-186°
Beilstein Reference: 154601
Formula:
C₁₀H₈N₂O₂ Punto di fusione: 185…186 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00041234 CAS Number: 7471-63-8 EINECS: 231-268-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...