Stampa…

5-Nitroquinoline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

N0344-25G N0344-250G
TCIAN0344-25GEA 74 EUR
TCIAN0344-25G TCIAN0344-250G
5-Nitroquinoline ≥98.0% (by GC, titration analysis)
5-Nitroquinoline
Formula: C₉H₆N₂O₂
Peso molecolare: 174,16 g/mol
Punto di ebollizione: 323 °C (1013 hPa)
Punto di fusione: 71 °C
MDL Number: MFCD00006790
CAS Number: 607-34-1
EINECS: 210-134-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5814b7cce058dc79bcfc705cc9551fdd15a29ed&vid=3f681aec818c7d2925dd6519fb90751cd919bbf7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 70.0 to 73.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR