5-Quinolinecarboxylic acid ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97+%
CAS No.: 7250-53-5
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point:
Formula:
C₁₀H₇NO₂ Punto di fusione: >300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00047608 CAS Number: 7250-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...