4-Cyclohexylbenzoic acid ≥98%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 98%
CAS No.: 20029-52-1
Molecular Formula: C13H16O2
Formula Weight: 204.27
Melting Point: 194-198°
Beilstein Reference: 1966671
Formula:
C₁₃H₁₆O₂ Punto di fusione: 194…198 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00039459 CAS Number: 20029-52-1 EINECS: 243-472-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...