Stampa…

Benzyldimethylhexadecylammonium chloride ≥97.0% (by HPLC)

Fornitore: TCI
Attenzione

B0237-25G B0237-500G
TCIAB0237-25GEA 40 EUR
TCIAB0237-25G TCIAB0237-500G
Benzyldimethylhexadecylammonium chloride ≥97.0% (by HPLC)
Benzyldimethylhexadecylammonium chloride
Formula: C₂₅H₄₆ClN
MDL Number: MFCD00011625
CAS Number: 122-18-9
EINECS: 204-526-3
Merck Index: 14,02017
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8348a129b257d87e0d6ba485db95e39de2e44b8e&vid=e185b133dbff435a1610fb4b384e02e2dfae8577&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (TPBNa Method) min. 95.0% (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR