Stampa…

2-Aminobenzenesulphonic acid ≥99.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

A0266-25G A0266-100G
TCIAA0266-25GEA 19 EUR
TCIAA0266-25G TCIAA0266-100G
2-Aminobenzenesulphonic acid ≥99.0% (by HPLC, titration analysis)
2-Aminobenzenesulphonic acid
Formula: C₆H₇NO₃S
MDL Number: MFCD00007705
CAS Number: 88-21-1
EINECS: 201-810-9
Merck Index: 14,06882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=571d23194b6b4dc5ccf529d56c9cc30ec1cdbbb4&vid=571d23194b6b4dc5ccf529d56c9cc30ec1cdbbb4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR