β-Phenylpropiophenone ≥98%
Fornitore: Thermo Scientific
Synonyms:
beta-Phenylpropiophenone, 1,3-Diphenyl-1-propanone
L09173.09
L09173.18
L09173.09EA
110
EUR
L09173.09
L09173.18
β-Phenylpropiophenone ≥98%
β-Phenylpropiophenone
Purity: 98%
CAS No.: 1083-30-3
Molecular Formula: C15H14O
Formula Weight: 210.28
Melting Point: 68-73°
Beilstein Reference: 1910661
Formula:
C₁₅H₁₄O Peso molecolare: 210,28 g/mol Punto di fusione: 68…73 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00039563 CAS Number: 1083-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...