4-Methoxybenzamidoxime ≥97%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
N-Hydroxy-4-methoxy-benzamidine
H52141.03
H52141.06
H52141.03EA
80.5
EUR
H52141.03
H52141.06
4-Methoxybenzamidoxime ≥97%
4-Methoxybenzamidoxime
Purity: 97%
CAS No.: 5373-87-5
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
MDL No.: MFCD05664434
Formula:
C₈H₁₀N₂O₂ Peso molecolare: 166,18 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00816993 CAS Number: 5373-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...