Methyldiphenylamine 97%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
N-Methyl-N-phenylaniline
414210050
414210250
ACRO414210050EA
45.1
EUR
ACRO414210050
ACRO414210250
Methyldiphenylamine 97%
Methyldiphenylamine
Formula:
C₁₃H₁₃N Peso molecolare: 183,25 g/mol Punto di fusione: –7,6 °C Flash Pt: >93 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00041900 CAS Number: 552-82-9 EINECS: 209-023-2 |
Risultati del test di specificazione
Appearance | Clear yellow to orange-red Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6215 to 1.6235 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...