Stampa…

Acido-3-amino-benzensolfonico 98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 1-Aminobenzene-3-sulphonic acid, Metanilic acid, 3-Aminobenzenesulphonic acid

413711000 413712500 413710010
ACRO413711000EA 24 EUR
ACRO413711000 ACRO413712500 ACRO413710010
Acido-3-amino-benzensolfonico 98%
Acido-3-amino-benzensolfonico
Formula: C₆H₇NO₃S
Peso molecolare: 173,19 g/mol
Punto di fusione: >300 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00065345
CAS Number: 121-47-1
EINECS: 204-473-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to grey-beige Powder
Infrared spectrum Conforms
Additional info product may have a purple-pink cast
Titration with NaOH ≥97.5 %
Stabilizer About 0.05% 3,5-di-tert-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR